Order Entry
Germany
ContactUsLinkComponent
Safety protection kit, PPE
Safety protection kit, PPE
Catalog # 111-2226
Supplier:  Portwest
undefined
Safety protection kit, PPE
Catalog # 111-2226
Supplier:  Portwest

Specifications

  • Colour:
    Yellow
  • Description:
    PPE protection kit, includes PC visor, PP brow guard and ABS earmuffs
  • L×W:
    400x200 mm
  • Pk:
    1 KIT

Specifications

About this item

The kit contains one pair of earmuffs and one clear visor with mount. It protects from particles, metal splashes and excessive noise.

  • Pressure-seal ear protector cup for optimum attenuation: SNR 25 dB
  • Polycarbonate visor with optical quality (1) and grade B impact resistance: 400×200 mm
  • Premium ratchet-tight adjustable suspension system for a snug fit

Colour: Yellow

Certifications: EN 166 1B (visor), EN 166 3B (brow guard)