Order Entry
Germany
ContactUsLinkComponent
Transport boxes, Biotainer
Transport boxes, Biotainer
Catalog # CORT3BIO000924
Supplier:  E3 Cortex
Transport boxes, Biotainer
Catalog # CORT3BIO000924
Supplier:  E3 Cortex

Specifications

  • Description:
    Transport box for hazardous material, load capacity 7 kg
  • Seq:
    02
  • W×D×H:
    270×260×370 mm
  • Pk:
    1

Specifications

About this item

These triple packaging designed for transporting all types of category A hazardous materials (UN 2814 / UN 2900 – class 6.2) at room temperature.

Certifications: Complies with regulation in force (IATA, IMDG, RID, ADR).