Cell analysers, Cellometer® Vision
The Cellometer Vision cell analysers are dedicated to cell analyses in toxicology, diabetes and obesity diseases, and for cell therapy research.
- Flexibility from ultraviolet to infrared optics for GFP, RFP, DAPI and Hoechst
- Maintenance free system thanks to disposable counting chambers
- Robust optical modules and light source
Fast results of call images, counts, measurements and viability calculations, pre-set protocols and cell types are combined with dual fluorescence for dual staining analysis of primary cells in heterogenous samples like blood making these equipments performant and reliable.
Cell imaging allow to check morphology and export images for presentations, pattern recognition software, automated data management.
: GMP and GLP compatible