Pipette tips, multi crystal
Catalog # SORE17000
Supplier: Sorenson Bioscience
Specifications
- Packed:Bulk (1000/bag)
- Volume:0,5 - 10 µl
- Seq:01
- Pk:1.000
Specifications
About this item
Virgin PP, non sterile, transparent.
- Conical top and smooth narrow lumen for maximum sample recovery and accuracy
- Manufactured from virgin materials and guaranteed free from pyrogens and heavy metals
- Certified free from RNases, DNases, human DNA and PCR inhibitors
These 10 μl pipette tips are optimised to fit Eppendorf pipettes.