ContactUsLinkComponent
Gas regulators
Gas regulators
Supplier:  Agilent
Certificates
Gas regulators
Supplier:  Agilent
Specifications
  • Catalog No:
  • HEWL0101-1536
  • HEWL5188-5374
  • HEWL0101-1399
  • HEWL0101-1398
  • HEWL0101-1400
  • HEWL0101-1535
  • HEWL0101-1537
  • Type:
  • Two stage
  • Two stage
  • Two stage
  • Two stage
  • Two stage
  • Two stage
  • Max. inlet pressure:
  • 207 bar
  • 207 bar
  • 207 bar
  • 207 bar
  • 207 bar
  • 207 bar
  • 207 bar
  • Outlet pressure:
  • 1,03 bar
  • 1,03 bar
  • 1,03 bar
  • 1,03 bar
  • 1,03 bar
  • 1,03 bar
Specifications