Order Entry
Germany
ContactUsLinkComponent
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 550) [clone: M2]
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 550) [clone: M2]
Catalog # COBSD9-1718
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 550) [clone: M2]
Catalog # COBSD9-1718

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    DYKDDDDK tag
  • Antigen symbol:
    DYKDDDDK
  • Clonality:
    Monoclonal
  • Clone:
    M2
  • Conjugation:
    DyLight® 550
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: