Anti-IgE Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-112-E
Supplier: Columbia Biosciences
Specifications
- Antibody type:Secondary
- Antigen symbol:IgE
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:Bovine, Rabbit
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Secondary
Antigen: IgE (Mouse) (mouse IgE)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse