Order Entry
Germany
ContactUsLinkComponent
Anti-V5 Tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # COBSD3-1834
Anti-V5 Tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # COBSD3-1834

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    V5 tag
  • Antigen symbol:
    V5
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: