Anti-T7 Tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # COBSD3-1832
Supplier: Columbia Biosciences
Specifications
- Antibody type:Tag
- Antigen name:T7 tag
- Antigen symbol:T7
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: