Order Entry
Germany
ContactUsLinkComponent
Anti-c-Myc tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]
Anti-c-Myc tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]
Catalog # COBSD3-1714-100
Anti-c-Myc tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]
Catalog # COBSD3-1714-100

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    v-myc avian myelocytomatosis viral oncogene homolog
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Clone:
    9E10
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    none
  • Gene ID:
    4609
  • Antigen synonyms:
    MRTL|bHLHe39|c-Myc|MYCC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: