Order Entry
Germany
ContactUsLinkComponent
Anti-Apolipoprotein A1 Goat Polyclonal Antibody
Anti-Apolipoprotein A1 Goat Polyclonal Antibody
Catalog # IMMRIR1371
Supplier:  ImmunoReagents
Anti-Apolipoprotein A1 Goat Polyclonal Antibody
Catalog # IMMRIR1371
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein A1
  • Antigen symbol:
    Apo-A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0.2 µm filtered
  • Antigen synonyms:
    Apolipoprotein A-I|apolipoprotein A1|Entrez:335|apo(a)
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human Apolipoprotein A1
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human Apolipoprotein A1
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Type: Primary
Antigen: Apolipoprotein A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human