Anti-V5 Tag Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIV0500-00T9C
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen name:V5 tag
- Antigen symbol:V5
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Form:liquid
- Storage buffer:25mM HEPES, pH 7.0, 0,05M sodium chloride,0,1% BSA, 0,005% thimerosal, 50% glycerol
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:1000-1:5000
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: