Order Entry
Germany
ContactUsLinkComponent
Anti-NCOA3 Mouse Monoclonal Antibody [clone: AC3]
Anti-NCOA3 Mouse Monoclonal Antibody [clone: AC3]
Catalog # USBIS6505-08G
Supplier:  US Biological
Anti-NCOA3 Mouse Monoclonal Antibody [clone: AC3]
Catalog # USBIS6505-08G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear receptor coactivator 3
  • Antigen symbol:
    NCOA3
  • Clonality:
    Monoclonal
  • Clone:
    AC3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: NCOA3 (nuclear receptor coactivator 3)
Clonality: Monoclonal
Clone: AC3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human