Anti-SIRT 5 Sheep Polyclonal Antibody (Biotin)
Catalog # USBIS1013-88J6
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 5
- Antigen symbol:SIRT5
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:33 to 310
- Storage buffer:PBS, 50ug/1ug BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: SIRT 5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human