Anti-Succinate Dehydrogenase Complex Assembly Factor 1, SDHAF1 Goat Polyclonal Antibody
Catalog # USBIS0553-27
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Succinate dehydrogenase complex assembly factor 1
- Antigen symbol:SDHAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, pH 7,2, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-HDSTGAPEWDGR
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by peptide affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml
Type: Primary
Antigen: Succinate Dehydrogenase Complex Assembly Factor 1, SDHAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human