Anti-PSMB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIP9078-38E-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) Subunit, Beta type 1
- Antigen symbol:PSMB1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:212 to 241
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Flow Cytometry: 1:10-1:50; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500
Type: Primary
Antigen: PSMB1 (proteasome (prosome, macropain) subunit, beta type, 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse