Anti-POU Domain, Class 2, Transcription Factor 2 Mouse Monoclonal Antibody [clone: 3E3-6F9]
USBIP5250-05C
- Antibody type:Primary
- Antigen name:POU Domain, Class 2, Transcription Factor 2
- Clonality:Monoclonal
- Clone:3E3-6F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 401
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: POU Domain, Class 2, Transcription Factor 2
Clonality: monoclonal
Clone: 3E3-6F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human