Anti-Peroxidase Anti-Peroxidase Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIP3350-36
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Peroxidase Anti-Peroxidase
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Sheep
- Isotype:IgG
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 0,01% thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Peroxidase Anti-Peroxidase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: