Anti-NIFK Mouse Monoclonal Antibody [clone: 9A39]
Catalog # USBIN2566-75D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nucleolar Protein Interacting With The Fha Domain Of Mki67
- Antigen symbol:NIFK
- Clonality:Monoclonal
- Clone:9A39
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,2% gelatin, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NIFK (nucleolar protein interacting with the FHA domain of MKI67)
Clonality: monoclonal
Clone: 9A39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human