Anti-Melanin Concentrating Hormone Receptor 1 Rabbit Polyclonal Antibody
Catalog # USBIM2867-75H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Melanin Concentrating Hormone Receptor 1
- Antigen symbol:MCHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10-18?g/ml
Type: Primary
Antigen: Melanin Concentrating Hormone Receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human