Order Entry
Germany
ContactUsLinkComponent
Anti-Matrix Metalloproteinase 9 Mouse Monoclonal Antibody [clone: 10F166]
Anti-Matrix Metalloproteinase 9 Mouse Monoclonal Antibody [clone: 10F166]
Artikelnr. USBIM2425-01H
Supplier:  US Biological
Anti-Matrix Metalloproteinase 9 Mouse Monoclonal Antibody [clone: 10F166]
Artikelnr. USBIM2425-01H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 9
  • Antigen symbol:
    MMP9
  • Clonality:
    Monoclonal
  • Clone:
    10F166
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: Matrix Metalloproteinase 9
Clonality: monoclonal
Clone: 10F166
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human