Anti-Histone H3, acetylated Rabbit polyclonal antibody
USBIH5110-12M7
- Antibody type:Primary
- Antigen name:Histone H3
- Antigen symbol:H3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Modification:acetylated
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM NaCl, 100ug/ml BSA and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Chromatin
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:25 ChImmunoprecipitation: 1:25
Type: Primary
Antigen: Histone H3, acetylated
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat