Order Entry
Germany
ContactUsLinkComponent
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 11H6]
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 11H6]
  USBIG8900-07A
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 11H6]
  USBIG8900-07A

 

  • Antibody type:
    Primary
  • Antigen name:
    Gram Negative Endotoxin
  • Clonality:
    Monoclonal
  • Clone:
    11H6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Gram Negative Endotoxin
Clonality: monoclonal
Clone: 11H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: