Anti-Fibrin, Fibrinogen Mouse Monoclonal Antibody [clone: MFB-HB]
Catalog # USBIF4197-37
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibrin, Fibrinogen
- Clonality:Monoclonal
- Clone:MFB-HB
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human,Dog
- Form:liquid
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:30-1:50
Type: Primary
Antigen: Fibrin, Fibrinogen
Clonality: monoclonal
Clone: MFB-HB
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog, Human