Anti-v-Ets Erythroblastosis Virus E26 Oncogene Homolog 1 Mouse Monoclonal Antibody [clone: 8C132]
USBIE8850-75
- Antibody type:Primary
- Antigen name:v-Ets Erythroblastosis Virus E26 Oncogene Homolog 1
- Clonality:Monoclonal
- Clone:8C132
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: v-Ets Erythroblastosis Virus E26 Oncogene Homolog 1
Clonality: monoclonal
Clone: 8C132
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: