Order Entry
Germany
ContactUsLinkComponent
Anti-DOCK2 Rabbit Polyclonal Antibody
Anti-DOCK2 Rabbit Polyclonal Antibody
  USBID8080-20B
Anti-DOCK2 Rabbit Polyclonal Antibody
  USBID8080-20B

 

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 2
  • Antigen symbol:
    DOCK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Horse,
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 0.5mg/ml

Type: Primary
Antigen: DOCK2 (dedicator of cytokinesis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse, Human, Mouse