Anti-DOCK1 Goat Polyclonal Antibody
Catalog # USBID8080-07D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 1
- Antigen symbol:DOCK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-TTRKQTSVDSGIVQ
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 3-4?g/ml
Type: Primary
Antigen: DOCK1 (dedicator of cytokinesis 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat