Anti-Digoxigenin Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
USBID8029-50H2
- Antibody type:Tag
- Antigen name:Digoxigenin
- Antigen symbol:DIG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Chicken
- ImmunoPrecipitation:Yes
- Isotype:IgY
- Western blot:Yes
- Form:liquid
- Storage buffer:25mM HEPES-saline, 0,1% BSA, 0,001% thimerosal, 50% glycerol
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:5000; Western Blot: 1:500
Type: Tag
Antigen: Digoxigenin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: