Anti-Digoxigenin Sheep polyclonal antibody
Catalog # USBID8029-50C
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen name:Digoxigenin
- Antigen symbol:DIG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: ELISA: 1:100-1:5000
Type: Tag
Antigen: Digoxigenin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: