Order Entry
Germany
ContactUsLinkComponent
Anti-Cysteine and Glycine-rich Protein 3 Chicken Polyclonal Antibody
Anti-Cysteine and Glycine-rich Protein 3 Chicken Polyclonal Antibody
Catalog # USBIC9001-21
Supplier:  US Biological
Anti-Cysteine and Glycine-rich Protein 3 Chicken Polyclonal Antibody
Catalog # USBIC9001-21
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cysteine and Glycine-rich Protein 3
  • Antigen symbol:
    CSRP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Cysteine and Glycine-rich Protein 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat