Order Entry
Germany
ContactUsLinkComponent
Anti-Cyclic AMP Rabbit Polyclonal Antibody
Anti-Cyclic AMP Rabbit Polyclonal Antibody
Catalog # USBIC8450-05A
Supplier:  US Biological
Anti-Cyclic AMP Rabbit Polyclonal Antibody
Catalog # USBIC8450-05A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic Adenosine Monophosphate
  • Antigen symbol:
    cAMP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Cyclic AMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: