Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-1]
Catalog # USBIC7554-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Colon Carcinoma
- Clonality:Monoclonal
- Clone:CC-1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Format:Supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:10 Stains acetone or ethanol fixed tissues
Type: Primary
Antigen: Colon Carcinoma
Clonality: Monoclonal
Clone: CC-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: