Anti-Citrate Lyase Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIC5699-50B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Citrate Lyase
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2 1ml sterile ddH2O. Dilute further
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation and ion exchange chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Citrate Lyase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: