Anti-cdk1, p34cdc2 Mouse monoclonal antibody [clone: 5F291]
USBIC2571-21A
- Antibody type:Primary
- Antigen symbol:cdk1, p34cdc2
- Clonality:Monoclonal
- Clone:5F291
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:20-1:40
Type: Primary
Antigen: cdk1, p34cdc2
Clonality: Monoclonal
Clone: 5F291
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human