Anti-CDK1 Mouse monoclonal antibody [clone: POH1]
USBIC2571-07H
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Monoclonal
- Clone:POH1
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: POH1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human