Anti-CDK1 Rabbit polyclonal antibody
Catalog # USBIC2549-49Q
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot:1:500-1:1000
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat