Anti-CDK1 Rabbit polyclonal antibody
100 µl
Catalog # USBIC2549-48
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Monkey
Documents
Specifications
- Size:100 µl
- Western blot:Yes
- Clonality:Polyclonal
- Modification:phospho-Thr14
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Cyclin-dependent kinase 1
- Cat. No.:USBIC2549-48
- Reactivity:Hamster|Monkey|Human
- Conjugation:Unconjugated
- Antigen symbol:CDK1
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol, 0,02% sodium azide.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



