Anti-CD51/CD61 Integrin alphaV beta3 Mouse Monoclonal Antibody [clone: 4j22]
Catalog # USBIC2405-17A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin alphaV beta3
- Antigen symbol:CD51/CD61
- Clonality:Monoclonal
- Clone:4j22
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:40-1:80
Type: Primary
Antigen: CD51/CD61 Integrin alphaV beta3
Clonality: Monoclonal
Clone: 4j22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human