Anti-Bactericidal Permeability Increasing Proetin Mouse monoclonal antibody unconjugated [clone: 4E3]
USBIB0003-40C
- Antibody type:Primary
- Antigen name:Bactericidal Permeability Increasing Proetin
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Neutralisation:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% BSA.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: Bactericidal Permeability Increasing Proetin
Clonality: monoclonal
Clone: 4000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human