Anti-ADAM10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9H28]
Catalog # USBIA0859-20D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 10
- Antigen symbol:ADAM10
- Clonality:Monoclonal
- Clone:9H28
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Form:liquid
- Storage buffer:saline, containing up to 0,5% BSA and 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: ADAM10 (ADAM metallopeptidase domain 10)
Clonality: monoclonal
Clone: 9H28
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: