Anti-ACTA2 Rabbit Polyclonal Antibody AMCA
Catalog # USBIA0760-26K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Actin, alpha 2, smooth muscle, aorta
- Antigen symbol:ACTA2
- Clonality:Polyclonal
- Conjugation:AMCA (7-amino-4-methylcoumarin-3-acetic acid)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMphosphate, 150mM sodium chloride, pH 7,2, BSA, 2mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: polyclonal
Clone:
Conjugation: AMCA
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse