Anti-GluA1/GluR1 Glutamate Receptor Mouse Monoclonal Antibody [clone: S355-1]
Catalog # USBI167589
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:GluR1 Glutamate Receptor
- Antigen symbol:GLUA1
- Clonality:Monoclonal
- Clone:S355-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: GluA1/GluR1 Glutamate Receptor
Clonality: monoclonal
Clone: S355-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat