Anti-CDK1 Mouse Monoclonal Antibody [clone: 13k26]
USBI143153
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Monoclonal
- Clone:13k26
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM NaCl, 0,2% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:100; Immunochemistry: 1:50
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: monoclonal
Clone: 13k26
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human