Anti-MYL2 Mouse Monoclonal Antibody [clone: 13H322]
USBI138168
- Antibody type:Primary
- Antigen name:Myosin Light Chain 2, regulatory, cardiac, slow
- Antigen symbol:MYL2
- Clonality:Monoclonal
- Clone:13H322
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 166
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: MYL2 (myosin, light chain 2, regulatory, cardiac, slow)
Clonality: monoclonal
Clone: 13H322
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat