Order Entry
Germany
ContactUsLinkComponent
Anti-CALB1 Mouse Monoclonal Antibody [clone: 8A11]
Anti-CALB1 Mouse Monoclonal Antibody [clone: 8A11]
Catalog # USBI136868
Supplier:  US Biological
Anti-CALB1 Mouse Monoclonal Antibody [clone: 8A11]
Catalog # USBI136868
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    calbindin 1, 28kDa
  • Antigen symbol:
    CALB1
  • Clonality:
    Monoclonal
  • Clone:
    8A11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgM kappa
  • Reactivity:
    Human
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Immunochemistry: 1:25-1:50

Type: Primary
Antigen: CALB1 (calbindin 1, 28kDa)
Clonality: monoclonal
Clone: 8A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human