Anti-CDK1 Mouse Monoclonal Antibody
USBI135970
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µl
- Pk:50 µl
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:200
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat