Anti-MPZL2 Mouse Monoclonal Antibody [clone: 2E10]
Catalog # USBI129836
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:myelin Protein zero-like 2
- Antigen symbol:MPZL2
- Clonality:Monoclonal
- Clone:2E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:27 to 136
- Storage buffer:PBS, pH 7,2.
- Sequence:VEIYTSRVLEAVNGTDARLKCTFSSFAPVGDALTVTWNFRPLDGGPEQFVFYYHIDPFQPMSGRFKDRVSWDGNPERYDASILLWKLQFDDNGTYTCQVKNPPDVDGVIG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MPZL2 (myelin protein zero-like 2)
Clonality: monoclonal
Clone: 20000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human