Anti-HOXC5 Mouse Monoclonal Antibody [clone: 1E10]
USBI128035
- Antibody type:Primary
- Antigen name:homeobox C5
- Antigen symbol:HOXC5
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 68
- Storage buffer:PBS, pH 7,2.
- Sequence:MSSYVANSFYKQSPNIPAYNMQTCGNYGSASEVQASRYCYGGLDLSITFPPPAPSNSLHGVDMAANPR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HOXC5 (homeobox C5)
Clonality: monoclonal
Clone: 10000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human