Anti-AP3B1 Mouse Monoclonal Antibody [clone: 3B4]
Catalog # USBI123381
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 3, beta 1 subunit
- Antigen symbol:AP3B1
- Clonality:Monoclonal
- Clone:3B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:995 to 1095
- Storage buffer:PBS, pH 7,2.
- Sequence:KEQGVLTGMNETSAVIIAAPQNFTPSVIFQKVVNVANVGAVPSGQDNIHRFAAKTVHSGSLMLVTVELKEGSTAQLIINTEKTVIGSVLLRELKPVLSQG*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: AP3B1 (adaptor-related protein complex 3, beta 1 subunit)
Clonality: monoclonal
Clone: 3B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human