Anti-VSV-G Tag Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI043789-PE
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen name:VSV-G tag
- Antigen symbol:VSV-G
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:5000; Western Blot: 1:50-100
Do not freeze!
Type: Tag
Antigen: VSV-G Tag (VSV-G tag)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: